Assay Details
Gene Symbol: GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020474.3 | NP_065207.2 | 1 - 2 | 234 | 103 | |
XM_005258239.3 | XP_005258296.1 | 2 - 3 | 302 | 103 | ||
XM_017025692.1 | XP_016881181.1 | 2 - 3 | 481 | 103 | ||
GenBank mRNA | AK293062.1 | - | 3 - 4 | 376 | 103 | |
BC047746.1 | - | 1 - 2 | 229 | 103 | ||
DN996147.1 | - | 1 - 2 | 252 | 103 | ||
U41514.1 | - | 1 - 2 | 171 | 103 | ||
X85018.1 | - | 1 - 2 | 140 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000145.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR18001644.hg.1 | ENST00000269195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001644.hg.1 | ENST00000537549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001644.hg.1 | ENST00000537748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001644.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001644.hg.1 | TC18000145.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001641.hg.1 | ENST00000269195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001641.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001641.hg.1 | TC18000145.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18001646.hg.1 | ENST00000269195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001646.hg.1 | ENST00000537549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001646.hg.1 | ENST00000537748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001646.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001646.hg.1 | TC18000145.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | TC1800007080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148551.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | TC1800007080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148554.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | TC1800007080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148556.hg.1 | NM_020474.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148896 | - | - |
