Assay Details
Gene Symbol: GALNT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020474.3 | NP_065207.2 | 8 - 9 | 1396 | 83 | |
XM_005258239.3 | XP_005258296.1 | 9 - 10 | 1464 | 83 | ||
XM_017025692.1 | XP_016881181.1 | 9 - 10 | 1643 | 83 | ||
GenBank mRNA | AJ505952.1 | - | 1 - 2 | 135 | 83 | |
BC038440.1 | - | 9 - 10 | 1461 | 4478643 | 83 | |
X85018.1 | - | 8 - 9 | 1302 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000145.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001654.hg.1 | ENST00000269195.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001654.hg.1 | ENST00000537549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001654.hg.1 | ENST00000537748.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18001654.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18001654.hg.1 | TC18000145.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | TC1800007080.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800148567.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000269195.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000586725.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000589189.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000590654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591081.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | ENST00000591924.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007080.hg.1 | NM_020474.3 | Refseq |
