Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000378460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000526569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000528071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000530326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
NM_015231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000378460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000526569.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000528071.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
ENST00000530326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
NM_015231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181728.hg.1 |
TC1100010762.hg.1 |
Clariom D TC |