Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020526.hg.1 |
ENST00000378460.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020526.hg.1 |
ENST00000426372.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020526.hg.1 |
ENST00000530326.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020526.hg.1 |
NM_015231.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020526.hg.1 |
TC11001750.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000378460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000530326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000532773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
NM_015231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000378460.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000530326.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
ENST00000532773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
NM_015231.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181713.hg.1 |
TC1100010762.hg.1 |
Clariom D TC |