Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005003.hg.1 |
ENST00000373255.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005003.hg.1 |
ENST00000395046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005003.hg.1 |
ENST00000479444.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005003.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005003.hg.1 |
TC10000425.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005000.hg.1 |
ENST00000373255.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005000.hg.1 |
ENST00000395046.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005000.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10005000.hg.1 |
TC10000425.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153718.hg.1 |
TC1000007913.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000153717.hg.1 |
TC1000007913.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000373255.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
ENST00000455786.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000007913.hg.1 |
NM_018649.2 |
Refseq |