Assay Details
Gene Symbol: HIST1H4G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003547.2 | NP_003538.1 | 1 | 316 | 68 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813735 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813727 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625488 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625462 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813734 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813729 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625470 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813737 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625469 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625485 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813719 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625452 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625459 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625460 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813736 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625484 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813718 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P04625461 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P24813742 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 10778 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 20157 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P277673 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P277673 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 15660 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P136386 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P136386 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 22004 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P17486016 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P277673 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_24_P277673 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00804713 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813737 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625462 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625486 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813719 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625468 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625452 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813735 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813720 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813727 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625486 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813741 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625466 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625469 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625468 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813738 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625484 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813719 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813718 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813737 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813739 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813742 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625488 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625485 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625460 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813744 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P24813721 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P04625462 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P277673 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P136386 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625456 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625462 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625485 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625460 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625476 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625457 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625478 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625487 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625471 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625452 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625453 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625475 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625466 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625455 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625480 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625486 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625469 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625464 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P04625483 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625471 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625469 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P15793762 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625470 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P04625472 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P136386 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P136386 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P277673 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 4313 | - | - |
Custom Human Agilent Array 11k, Chromosome 6, Expression | 9902 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 128751 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 28724 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P136386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625521 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625472 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625455 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625456 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625480 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625485 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625439 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625425 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625453 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793755 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625460 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625491 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625518 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15793781 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625514 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625510 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625517 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625417 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625464 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625478 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625501 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625435 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625429 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625448 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625493 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625507 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625421 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625487 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625490 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625444 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625505 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625446 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625433 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625469 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625513 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625431 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625471 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625483 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625462 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625475 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625508 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625466 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P04625427 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P136386 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_003547_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124436 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124434 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8124435 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8124433 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208551_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738789_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11738789_at | - | - |
Human Genome U133 Plus 2.0 Array | 208551_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 208551_at | - | - |
Human Genome U133A 2.0 Array | 208551_PM_at | - | - |
Human Genome U133A 2.0 Array | 208551_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_003547_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw533469 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208551_at6 | - | - |
Human Genome U133 Set | 208551_at | - | - |
Human Genome U133 Set | 208551_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029537 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A029537 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 178056 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1712808 | BC126276.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21166393-S | NM_003547.2 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_928-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0005340148 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0005340148 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 4890113 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_928 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1712808 | BC126278.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1712808 | NM_003547.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1712808 | BC126276.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1712808 | BC126278.1 | human_transcriptome |
