Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
TC01000781.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
ENST00000394993.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
ENST00000394994.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012459.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012453.hg.1 |
TC01000781.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012453.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012453.hg.1 |
ENST00000394994.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012453.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01012453.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000464140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100008779.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
ENST00000621530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162606.hg.1 |
TC0100008779.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
ENST00000328299.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
NM_001160011.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
NM_152996.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100162602.hg.1 |
TC0100008779.hg.1 |
Clariom D TC |