Assay Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305173.1 | NP_001292102.1 | 1 - 2 | 158 | 72 | |
| GenBank mRNA | AK058068.1 | - | 1 - 2 | 103 | 72 | |
| BC057843.1 | - | 1 - 2 | 150 | 5296090 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK058068.1, BC057843.1, NM_001305173.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180595.hg.1 | TC1600010480.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000563336.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600010480.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180599.hg.1 | TC1600010480.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000219301.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000543437.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000567164.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000569079.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | ENST00000569727.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001080492.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001305173.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | NM_001305174.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600180597.hg.1 | TC1600010480.hg.1 | Clariom D TC |

PRSS54
Genomic Map