Assay Details
| Gene Symbol: CYP2C19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000769.2 | NP_000760.1 | 4 - 5 | 668 | 169 | |
| GenBank mRNA | M61854.1 | - | 4 - 5 | 648 | 169 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10002971.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
xenobiotic metabolic process
steroid metabolic process
monoterpenoid metabolic process
drug metabolic process
epoxygenase P450 pathway
exogenous drug catabolic process
heterocycle metabolic process
oxidation-reduction process
omega-hydroxylase P450 pathway
monooxygenase activity
iron ion binding
arachidonic acid epoxygenase activity
steroid hydroxylase activity
oxidoreductase activity
(S)-limonene 6-monooxygenase activity
(S)-limonene 7-monooxygenase activity
oxygen binding
enzyme binding
heme binding
(R)-limonene 6-monooxygenase activity
xenobiotic metabolic process
steroid metabolic process
monoterpenoid metabolic process
drug metabolic process
epoxygenase P450 pathway
exogenous drug catabolic process
heterocycle metabolic process
oxidation-reduction process
omega-hydroxylase P450 pathway
monooxygenase activity
iron ion binding
arachidonic acid epoxygenase activity
steroid hydroxylase activity
oxidoreductase activity
(S)-limonene 6-monooxygenase activity
(S)-limonene 7-monooxygenase activity
oxygen binding
enzyme binding
heme binding
(R)-limonene 6-monooxygenase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10007718.hg.1 | TC10002971.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007718.hg.1 | ENST00000371321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007718.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007716.hg.1 | TC10002971.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007716.hg.1 | ENST00000371321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007716.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007717.hg.1 | TC10002971.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007717.hg.1 | ENST00000371321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007717.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011477.hg.1 | ENST00000457790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011477.hg.1 | ENST00000457790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007721.hg.1 | TC10002971.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007721.hg.1 | ENST00000371321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007721.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007720.hg.1 | TC10002971.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007720.hg.1 | ENST00000371321.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007720.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185958.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185960.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000011477.hg.1 | ENST00000457790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011477.hg.1 | ENST00000457790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185957.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000185962.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185962.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185962.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185962.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185962.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000185963.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185963.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185963.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185963.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185963.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000185959.hg.1 | TC1000012481.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000371321.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | ENST00000480405.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000012481.hg.1 | NM_000769.2 | Refseq |

CYP2C19
Genomic Map