Assay Details
Gene Symbol: C1RL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297640.1 | NP_001284569.1 | 2 - 3 | 399 | 75 | |
NM_001297642.1 | NP_001284571.1 | 2 - 3 | 399 | 75 | ||
NM_001297643.1 | NP_001284572.1 | 2 - 3 | 399 | 75 | ||
NM_016546.3 | NP_057630.2 | 2 - 3 | 399 | 75 | ||
XM_017019397.1 | XP_016874886.1 | 2 - 3 | 446 | 75 | ||
XM_017019398.1 | XP_016874887.1 | 2 - 3 | 446 | 75 | ||
XM_017019399.1 | XP_016874888.1 | 2 - 3 | 446 | 75 | ||
XM_017019400.1 | XP_016874889.1 | 2 - 3 | 446 | 75 | ||
XM_017019401.1 | XP_016874890.1 | 2 - 3 | 446 | 75 | ||
XM_017019402.1 | XP_016874891.1 | 2 - 3 | 446 | 75 | ||
XM_017019403.1 | XP_016874892.1 | 2 - 3 | 446 | 75 | ||
XM_017019404.1 | XP_016874893.1 | 2 - 3 | 446 | 75 | ||
GenBank mRNA | AF178985.1 | - | 2 - 3 | 318 | 75 | |
AK314299.1 | - | 2 - 3 | 75 | |||
BC062428.1 | - | 2 - 3 | 335 | 75 | ||
DB113812.1 | - | 2 - 3 | 394 | 75 | ||
DR004214.1 | - | 2 - 3 | 325 | 75 | ||
HY062376.1 | - | 2 - 3 | 399 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
