| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
TC07001996.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
ENST00000316286.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
ENST00000451334.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
ENST00000538412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07030677.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700193522.hg.1 |
TC0700012970.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000316286.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000451334.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
ENST00000491431.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700012970.hg.1 |
NM_152411.3 |
Refseq |