Assay Details
| Gene Symbol: UNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080419.2 | NP_001073888.2 | 14 - 15 | 2063 | 61 | |
| XM_006722145.3 | XP_006722208.1 | 17 - 18 | 6408 | 61 | ||
| XM_006722148.2 | XP_006722211.1 | 15 - 16 | 2204 | 61 | ||
| XM_011525400.1 | XP_011523702.1 | 15 - 16 | 2132 | 61 | ||
| XM_011525401.2 | XP_011523703.1 | 9 - 10 | 1220 | 61 | ||
| XM_017025247.1 | XP_016880736.1 | 17 - 18 | 6444 | 61 | ||
| XM_017025248.1 | XP_016880737.1 | 16 - 17 | 6346 | 61 | ||
| XM_017025249.1 | XP_016880738.1 | 16 - 17 | 2489 | 61 | ||
| XM_017025250.1 | XP_016880739.1 | 15 - 16 | 2391 | 61 | ||
| GenBank mRNA | AB051540.1 | - | 14 - 15 | 2046 | 61 | |
| BC053362.1 | - | 14 - 15 | 2046 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000862.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17011248.hg.1 | ENST00000293218.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011248.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011248.hg.1 | TC17000862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167203.hg.1 | TC1700008923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |

UNK
Genomic Map