Assay Details
Gene Symbol: ZNF219 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006720164.3 | XP_006720227.1 | 1 - 2 | 84 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006720164.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
