Assay Details
| Gene Symbol: PRKAR2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321982.1 | NP_001308911.1 | 2 - 3 | 552 | 110 | |
| NM_001321983.1 | NP_001308912.1 | 2 - 3 | 552 | 110 | ||
| NM_004157.3 | NP_004148.1 | 2 - 3 | 552 | 110 | ||
| XM_005265315.3 | XP_005265372.1 | 2 - 3 | 537 | 110 | ||
| XM_011533942.2 | XP_011532244.1 | 2 - 3 | 537 | 110 | ||
| XM_011533943.1 | XP_011532245.1 | 2 - 3 | 537 | 110 | ||
| XM_017006815.1 | XP_016862304.1 | 2 - 3 | 537 | 110 | ||
| XM_017006816.1 | XP_016862305.1 | 2 - 3 | 537 | 110 | ||
| GenBank mRNA | AK293042.1 | - | 2 - 3 | 506 | 110 | |
| BC002763.2 | - | 2 - 3 | 530 | 3629579 | 110 | |
| BT007225.1 | - | 2 - 3 | 298 | 110 | ||
| CF994284.1 | - | 2 - 3 | 566 | 110 | ||
| KU178276.1 | - | 2 - 3 | 298 | 110 | ||
| KU178277.1 | - | 2 - 3 | 298 | 110 | ||
| X14968.1 | - | 2 - 3 | 487 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
blood coagulation
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
renal water homeostasis
blood coagulation
activation of protein kinase A activity
intracellular signal transduction
cellular response to glucagon stimulus
negative regulation of cAMP-dependent protein kinase activity
cAMP-dependent protein kinase inhibitor activity
protein binding
cAMP-dependent protein kinase regulator activity
cAMP binding
ubiquitin protein ligase binding
protein kinase A catalytic subunit binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000437821.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000438535.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | ENST00000457914.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011036.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000265563.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000296446.12 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000419216.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | ENST00000454963.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | NM_004157.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300181311.hg.1 | TC0300011036.hg.1 | Clariom D TC |

PRKAR2A
Genomic Map