Assay Details
| Gene Symbol: MYO5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000259.3 | NP_000250.3 | 25 - 26 | 3674 | 114 | |
| NM_001142495.1 | NP_001135967.1 | 25 - 26 | 3674 | 114 | ||
| XM_005254397.3 | XP_005254454.1 | 25 - 26 | 3506 | 114 | ||
| XM_011521606.2 | XP_011519908.2 | 25 - 26 | 3509 | 114 | ||
| XM_011521607.2 | XP_011519909.2 | 25 - 26 | 3509 | 114 | ||
| XM_011521608.2 | XP_011519910.2 | 25 - 26 | 3508 | 114 | ||
| XM_011521609.2 | XP_011519911.2 | 25 - 26 | 3508 | 114 | ||
| XM_011521610.2 | XP_011519912.2 | 25 - 26 | 3509 | 114 | ||
| XM_011521611.2 | XP_011519913.2 | 25 - 26 | 3508 | 114 | ||
| XM_011521612.2 | XP_011519914.2 | 25 - 26 | 3508 | 114 | ||
| XM_017022227.1 | XP_016877716.1 | 24 - 25 | 3401 | 114 | ||
| GenBank mRNA | AB209487.1 | - | 18 - 19 | 2287 | 114 | |
| AB209505.1 | - | 18 - 19 | 2287 | 114 | ||
| AB290180.1 | - | 25 - 26 | 3433 | 114 | ||
| S74799.1 | - | 2 - 3 | 251 | 114 | ||
| U90942.1 | - | 25 - 26 | 3680 | 114 | ||
| Y07759.1 | - | 25 - 26 | 3680 | 114 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001456.hg.1
Set Membership: |
|

MYO5A
Genomic Map