Assay Details
Gene Symbol: FAM216A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013300.2 | NP_037432.2 | 3 - 4 | 759 | 69 | |
XM_005253875.4 | XP_005253932.1 | 3 - 4 | 269 | 69 | ||
XM_011538246.2 | XP_011536548.1 | 3 - 4 | 287 | 69 | ||
GenBank mRNA | AK225731.1 | - | 3 - 4 | 750 | 69 | |
AK297684.1 | - | 3 - 4 | 269 | 69 | ||
AK308802.1 | - | 3 - 4 | 372 | 69 | ||
BC020967.1 | - | 3 - 4 | 575 | 3848037 | 69 | |
CR457001.1 | - | 3 - 4 | 310 | 69 | ||
U79274.1 | - | 3 - 4 | 726 | 69 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000857.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297684.1,XM_005253875.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | ENST00000377673.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | ENST00000538285.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12011181.hg.1 | TC12000857.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000547539.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200008843.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000377673.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000538285.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000546396.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000548449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | ENST00000548869.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | NM_013300.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200165543.hg.1 | TC1200008843.hg.1 | Clariom D TC |
