Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001184963.1 | NP_001171892.1 | 3 - 4 | 234 | 72 | |
| NM_173160.2 | NP_775183.1 | 3 - 4 | 370 | 72 | ||
| GenBank mRNA | AY358584.1 | - | 3 - 4 | 234 | 72 | |
| BC054876.1 | - | 3 - 4 | 376 | 4610529 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000272.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003119.hg.1 | ENST00000374451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003119.hg.1 | ENST00000458363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003119.hg.1 | ENST00000476166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003119.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10003119.hg.1 | TC10000272.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000007415.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150401.hg.1 | TC1000007415.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000476166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000479189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000480834.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | ENST00000616495.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | NM_173160.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000150403.hg.1 | TC1000007415.hg.1 | Clariom D TC |

FXYD4
Genomic Map