Assay Details
Gene Symbol: TCEB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145653.3 | NP_663628.2 | 1 | 1566 | 65 | |
GenBank mRNA | AB076840.1 | - | 1 | 1330 | 65 |
Gene Symbol: LOC107983955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017026108.1 | XP_016881597.1 | 1 | 904 | 65 |
Gene Symbol: TCEB3CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100817.1 | NP_001094287.1 | 2 | 1330 | 65 | |
GenBank mRNA | BC103912.1 | - | 2 | 1326 | 65 | |
BC103913.1 | - | 2 | 1331 | 40006027 | 65 |
Gene Symbol: TCEB3CL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242907.1 | NP_001229836.1 | 1 | 1566 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000488.hg.1, TC18000489.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P31285626 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10685029 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10685027 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P10685028 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P315910 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P315910 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 21155 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56786 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P315910 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P315910 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P315910 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P315910 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685025 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685028 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685027 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285631 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285626 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685024 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685029 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685025 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31285626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685027 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285631 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685029 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285626 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685025 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685027 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P31285624 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685028 | - | - |
Agilent Whole Chr array_18 19 21 | A_17_P10685024 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685029 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685025 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10685027 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31285626 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P315910 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10685029 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10685024 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10685025 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P10685027 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P315910 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685047 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065200 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685029 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685065 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685032 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065200 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065204 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685049 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685067 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685040 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065192 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685056 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685032 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685039 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685031 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685030 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685027 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685036 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685042 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065192 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685054 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685044 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685052 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685008 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685059 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685029 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685027 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065214 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P10685025 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P17065196 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8023158 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023153 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023155 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023155 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023153 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023157 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023157 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023157 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023153 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8023155 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023154 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023152 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023154 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023156 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023152 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023152 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023154 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023156 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8023156 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1552860_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR18005098.hg.1 | ENST00000451265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005098.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005098.hg.1 | TC18000489.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR18005099.hg.1 | ENST00000451265.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005099.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005099.hg.1 | TC18000489.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008607.hg.1 | ENST00000620881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008607.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008607.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005095.hg.1 | NM_001100817.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005095.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005095.hg.1 | TC18000488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800008609.hg.1 | ENST00000611323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800008609.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800008609.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800009278.hg.1 | ENST00000620688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009278.hg.1 | ENST00000620688.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18005096.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18005096.hg.1 | TC18000488.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1800009279.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009279.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800009279.hg.1 | NM_001100817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158851.hg.1 | ENST00000611323.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800158851.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158851.hg.1 | TC1800008609.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800158851.hg.1 | ENST00000611323.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800158851.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008609.hg.1 | ENST00000611323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008609.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008609.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009279.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009279.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009279.hg.1 | NM_001100817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158843.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158843.hg.1 | TC1800008607.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800158843.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158852.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158852.hg.1 | TC1800008609.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800158852.hg.1 | NM_145653.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008607.hg.1 | ENST00000620881.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800008607.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1800008607.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158842.hg.1 | ENST00000620881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800158842.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800158842.hg.1 | TC1800008607.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800158842.hg.1 | ENST00000620881.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800158842.hg.1 | NM_001242907.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800164091.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800164091.hg.1 | NM_001100817.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800164091.hg.1 | TC1800009279.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800164091.hg.1 | ENST00000330682.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800164091.hg.1 | NM_001100817.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1800009278.hg.1 | ENST00000620688.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800009278.hg.1 | ENST00000620688.2 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1552860_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 1552860_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1552860_at6 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010297 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010296 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 168066 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 169670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1810590 | AB076840.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237069 | NM_001242907.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237069 | NM_001100817.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237069 | BC103912.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3237069 | BC103913.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237069 | NM_001242907.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237069 | NM_001100817.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237069 | BC103912.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3237069 | BC103913.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237069 | NM_001242907.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237069 | NM_001100817.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237069 | BC103912.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3237069 | BC103913.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237069 | NM_001242907.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237069 | NM_001100817.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237069 | BC103912.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3237069 | BC103913.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21717823-S | NM_001242907.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21717823-S | NM_001100817.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21717823-S | BC103912.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21717823-S | BC103913.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_21717823-S | NM_145653.3 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_6870-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0002100044 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0002100044 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1690705 | NM_001242907.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1690705 | NM_001100817.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1690705 | BC103912.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1690705 | BC103913.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 3400543 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_6870 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690705 | NM_001242907.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690705 | NM_001100817.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690705 | BC103912.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690705 | BC103913.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1810590 | AB076840.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1810590 | AB076840.1 | human_transcriptome |
