Assay Details
Gene Symbol: KIF18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031217.3 | NP_112494.3 | 6 - 7 | 1077 | 115 | |
XM_017018379.1 | XP_016873868.1 | 6 - 7 | 1633 | 115 | ||
GenBank mRNA | AB062483.1 | - | 6 - 7 | 978 | 115 | |
AK093850.1 | - | 2 - 3 | 264 | 115 | ||
AK312576.1 | - | 6 - 7 | 1022 | 115 | ||
AL136819.1 | - | 6 - 7 | 1024 | 115 | ||
AY791349.1 | - | 6 - 7 | 1007 | 115 | ||
BC026090.1 | - | 6 - 7 | 1057 | 115 | ||
BC048347.2 | - | 5 - 6 | 942 | 5576803 | 115 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001517.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
microtubule depolymerization
sister chromatid cohesion
mitotic metaphase plate congression
male meiosis
protein transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of microtubule cytoskeleton organization
seminiferous tubule development
actin binding
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
ubiquitin binding
microtubule plus-end binding
tubulin-dependent ATPase activity
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
microtubule depolymerization
sister chromatid cohesion
mitotic metaphase plate congression
male meiosis
protein transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
regulation of microtubule cytoskeleton organization
seminiferous tubule development
actin binding
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
ubiquitin binding
microtubule plus-end binding
tubulin-dependent ATPase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018984.hg.1 | ENST00000263181.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11018984.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11018984.hg.1 | TC11001517.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000526288.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100010418.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178987.hg.1 | TC1100010418.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000263181.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000531047.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | ENST00000533466.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | NM_031217.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100178988.hg.1 | TC1100010418.hg.1 | Clariom D TC |
