Assay Details
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098577.2 | NP_001092047.1 | 4 - 5 | 435 | 119 | |
GenBank mRNA | AK297483.1 | - | 4 - 5 | 371 | 119 | |
BE564858.1 | - | 3 - 4 | 353 | 119 | ||
CN269912.1 | - | 3 - 4 | 315 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000613.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
RNA binding
structural constituent of ribosome
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02009683.hg.1 | TC02000613.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02009683.hg.1 | ENST00000409028.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02009683.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02009684.hg.1 | TC02000613.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02009684.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000264258.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409000.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409038.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409320.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409650.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000409733.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000419276.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000441435.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | ENST00000456292.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_000993.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008632.hg.1 | NM_001099693.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160277.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160277.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160277.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160277.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160277.hg.1 | TC0200008632.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200160278.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160278.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160278.hg.1 | ENST00000409028.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160278.hg.1 | NM_001098577.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160278.hg.1 | TC0200008632.hg.1 | Clariom D TC |
