Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016402.hg.1 |
ENST00000228862.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016402.hg.1 |
ENST00000298573.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016402.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12016402.hg.1 |
TC12001255.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200175295.hg.1 |
TC1200009967.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000228862.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000298573.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000536236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000539940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000541207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
ENST00000545864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009967.hg.1 |
NM_030640.2 |
Refseq |