Assay Details
| Gene Symbol: SMC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005445.3 | NP_005436.1 | 5 - 6 | 390 | 103 | |
| GenBank mRNA | AF020043.1 | - | 5 - 6 | 355 | 103 | |
| AK289771.1 | - | 5 - 6 | 386 | 103 | ||
| AK315845.1 | - | 5 - 6 | 389 | 103 | ||
| BC047324.1 | - | 5 - 6 | 285 | 5493263 | 103 | |
| CA439554.1 | - | 5 - 6 | 404 | 103 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000802.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of DNA replication
DNA repair
mitotic spindle organization
sister chromatid cohesion
mitotic sister chromatid cohesion
mitotic nuclear division
meiotic nuclear division
signal transduction
protein sumoylation
stem cell population maintenance
negative regulation of DNA endoreduplication
cell division
chromatin binding
microtubule motor activity
protein binding
ATP binding
mediator complex binding
dynein binding
protein heterodimerization activity
regulation of DNA replication
DNA repair
mitotic spindle organization
sister chromatid cohesion
mitotic sister chromatid cohesion
mitotic nuclear division
meiotic nuclear division
signal transduction
protein sumoylation
stem cell population maintenance
negative regulation of DNA endoreduplication
cell division
chromatin binding
microtubule motor activity
protein binding
ATP binding
mediator complex binding
dynein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009600.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009600.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009600.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10009600.hg.1 | TC10000802.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008896.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | ENST00000361804.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | ENST00000462899.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | NM_005445.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000160804.hg.1 | TC1000008896.hg.1 | Clariom D TC |

SMC3
Genomic Map