Assay Details
Gene Symbol: SH3RF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020870.3 | NP_065921.2 | 9 - 10 | 2105 | 123 | |
GenBank mRNA | AB040927.1 | - | 5 - 6 | 1020 | 123 | |
AK021429.1 | - | 9 - 10 | 1966 | 123 | ||
AK025255.1 | - | 5 - 6 | 937 | 123 | ||
BC033203.1 | - | 8 - 9 | 1652 | 123 | ||
BC041023.1 | - | 7 - 8 | 1288 | 123 | ||
BC053671.1 | - | 7 - 8 | 2447 | 6144754 | 123 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001724.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
protein ubiquitination
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of JNK cascade
negative regulation of extrinsic apoptotic signaling pathway
protein binding
zinc ion binding
ligase activity
apoptotic process
protein ubiquitination
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of JNK cascade
negative regulation of extrinsic apoptotic signaling pathway
protein binding
zinc ion binding
ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04023841.hg.1 | TC04001724.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04023841.hg.1 | ENST00000284637.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04023841.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04023841.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000502315.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000510806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400012378.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000284637.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000508685.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | ENST00000511421.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | NM_020870.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400184839.hg.1 | TC0400012378.hg.1 | Clariom D TC |
