Assay Details
| Gene Symbol: ST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018412.3 | NP_060882.2 | 9 - 10 | 1217 | 97 | |
| NM_021908.2 | NP_068708.1 | 10 - 11 | 1286 | 97 | ||
| GenBank mRNA | AF234882.1 | - | 9 - 10 | 1216 | 97 | |
| AF234883.1 | - | 10 - 11 | 1285 | 97 | ||
| AJ277291.1 | - | 8 - 9 | 852 | 97 | ||
| AK093212.1 | - | 10 - 11 | 1466 | 97 | ||
| AK292372.1 | - | 9 - 10 | 1043 | 97 | ||
| AK297510.1 | - | 10 - 11 | 1163 | 97 | ||
| AK297861.1 | - | 10 - 11 | 994 | 97 | ||
| AK298540.1 | - | 9 - 10 | 1039 | 97 | ||
| AY009152.1 | - | 10 - 11 | 1277 | 97 | ||
| AY009153.1 | - | 9 - 10 | 1208 | 97 | ||
| BC030954.1 | - | 10 - 11 | 1022 | 97 | ||
| BC075855.1 | - | 10 - 11 | 1286 | 5562032 | 97 | |
| CR936656.1 | - | 4 - 5 | 427 | 97 | ||
| KJ534947.1 | - | 10 - 11 | 1072 | 97 | ||
More Information
Set Membership: |
|

ST7
Genomic Map