Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
ENST00000357596.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
ENST00000378319.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
ENST00000488958.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
NM_001040443.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
NM_001040444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002360.hg.1 |
TC13000199.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000357596.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000378319.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000426879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000442195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000460489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000465045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000488605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000488958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000495157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000496612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000621822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
NM_001040443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
NM_001040444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000357596.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000378319.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000426879.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000442195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000460489.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000465045.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000488605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000488958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000495157.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000496612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
ENST00000621822.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
NM_001040443.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
NM_001040444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148601.hg.1 |
TC1300007167.hg.1 |
Clariom D TC |