Assay Details
| Gene Symbol: TAOK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252043.1 | NP_001238972.1 | 4 - 5 | 1120 | 84 | |
| NM_004783.3 | NP_004774.1 | 4 - 5 | 1120 | 84 | ||
| NM_016151.3 | NP_057235.2 | 4 - 5 | 1120 | 84 | ||
| XM_011545982.1 | XP_011544284.1 | 4 - 5 | 1346 | 84 | ||
| XM_011545983.2 | XP_011544285.1 | 4 - 5 | 1346 | 84 | ||
| XM_011545984.1 | XP_011544286.1 | 4 - 5 | 1346 | 84 | ||
| XM_011545985.1 | XP_011544287.1 | 4 - 5 | 1346 | 84 | ||
| XM_011545986.2 | XP_011544288.1 | 4 - 5 | 1346 | 84 | ||
| GenBank mRNA | AB020688.1 | - | 4 - 5 | 706 | 84 | |
| AF061943.1 | - | 4 - 5 | 1061 | 84 | ||
| AF263313.1 | - | 3 - 4 | 303 | 84 | ||
| AK291473.1 | - | 4 - 5 | 637 | 84 | ||
| BC015204.1 | - | 4 - 5 | 1086 | 84 | ||
| BC031825.2 | - | 4 - 5 | 378 | 84 | ||
| BC051798.1 | - | 4 - 5 | 856 | 84 | ||
| BC054495.1 | - | 4 - 5 | 1073 | 84 | ||
| BC136653.1 | - | 4 - 5 | 805 | 9020643 | 84 | |
| BC136655.1 | - | 4 - 5 | 805 | 9020645 | 84 | |
| BC142663.1 | - | 4 - 5 | 460 | 40148112 | 84 | |
| BC144344.1 | - | 4 - 5 | 805 | 9052864 | 84 | |
| BC151221.1 | - | 4 - 5 | 460 | 40148752 | 84 | |
| BC152413.1 | - | 4 - 5 | 706 | 8862518 | 84 | |
More Information
Set Membership: |
|

TAOK2
Genomic Map