Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286132.1 | NP_001273061.1 | 1 - 2 | 191 | 79 | |
NM_001286133.1 | NP_001273062.1 | 1 - 2 | 191 | 79 | ||
NM_006638.3 | NP_006629.2 | 1 - 2 | 191 | 79 | ||
XM_011514253.2 | XP_011512555.1 | 1 - 2 | 186 | 79 | ||
GenBank mRNA | BC017871.1 | - | 1 - 2 | 165 | 4278825 | 79 |
BG430780.1 | - | 1 - 2 | 166 | 79 | ||
BM015573.1 | - | 1 - 2 | 223 | 79 | ||
BM827175.1 | - | 1 - 2 | 139 | 79 | ||
BQ220155.1 | - | 1 - 2 | 139 | 79 | ||
BX951376.1 | - | 1 - 2 | 191 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06001239.hg
Additional Information:
This assay is in the 5'UTR region of the BC017871.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06014740.hg.1 | TC06001239.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06014740.hg.1 | ENST00000319533.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014740.hg.1 | ENST00000380051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014740.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014740.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06014742.hg.1 | TC06001239.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06014742.hg.1 | ENST00000319533.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014742.hg.1 | ENST00000380051.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06014742.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172593.hg.1 | TC0600010681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172591.hg.1 | TC0600010681.hg.1 | Clariom D TC |
