Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286132.1 | NP_001273061.1 | 2 - 3 | 332 | 151 | |
GenBank mRNA | BC017871.1 | - | 2 - 3 | 306 | 4278825 | 151 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC017871.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172585.hg.1 | TC0600010681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600172587.hg.1 | TC0600010681.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000319533.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000380051.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000464646.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000468105.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000479782.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000483546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | ENST00000618533.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286132.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_001286133.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600010681.hg.1 | NM_006638.3 | Refseq |
