Assay Details
| Gene Symbol: EYA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172110.3 | NP_742108.2 | 12 - 13 | 1573 | 77 | |
| GenBank mRNA | BC013882.2 | - | 12 - 13 | 1573 | 77 | |
More Information
Set Membership: |
|

EYA2
Genomic Map