Assay Details
| Gene Symbol: BLK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001715.2 | NP_001706.2 | 5 - 6 | 944 | 98 | |
| XM_011543824.1 | XP_011542126.1 | 5 - 6 | 965 | 98 | ||
| XM_011543825.2 | XP_011542127.1 | 5 - 6 | 710 | 98 | ||
| XM_011543827.1 | XP_011542129.1 | 4 - 5 | 841 | 98 | ||
| XM_011543828.2 | XP_011542130.1 | 5 - 6 | 965 | 98 | ||
| XM_011543829.2 | XP_011542131.1 | 5 - 6 | 965 | 98 | ||
| XM_017013730.1 | XP_016869219.1 | 4 - 5 | 841 | 98 | ||
| GenBank mRNA | AK313751.1 | - | 5 - 6 | 533 | 98 | |
| BC007371.2 | - | 5 - 6 | 507 | 3634568 | 98 | |
| BC018703.1 | - | 5 - 6 | 808 | 98 | ||
| BC032413.1 | - | 5 - 6 | 553 | 5216030 | 98 | |
| S76617.1 | - | 5 - 6 | 951 | 98 | ||
| Z33998.1 | - | 5 - 6 | 585 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000087.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
transmembrane receptor protein tyrosine kinase signaling pathway
cell differentiation
positive regulation of insulin secretion
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
negative regulation of apoptotic process
innate immune response
regulation of phagocytosis
B cell receptor signaling pathway
regulation of podosome assembly
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
cytoskeleton organization
transmembrane receptor protein tyrosine kinase signaling pathway
cell differentiation
positive regulation of insulin secretion
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
negative regulation of apoptotic process
innate immune response
regulation of phagocytosis
B cell receptor signaling pathway
regulation of podosome assembly
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000821.hg.1 | TC08000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000821.hg.1 | ENST00000259089.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000821.hg.1 | ENST00000427279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000821.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000821.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000820.hg.1 | TC08000087.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000820.hg.1 | ENST00000259089.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000820.hg.1 | ENST00000427279.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000820.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08000820.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145705.hg.1 | TC0800006746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145704.hg.1 | TC0800006746.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000259089.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000525389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000526778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000529894.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | ENST00000533828.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006746.hg.1 | NM_001715.2 | Refseq |

BLK
Genomic Map