Assay Details
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021642.3 | NP_067674.2 | 1 - 2 | 135 | 66 | |
XM_017000663.1 | XP_016856152.1 | 1 - 2 | 120 | 66 | ||
XM_017000668.1 | XP_016856157.1 | 1 - 2 | 120 | 66 | ||
GenBank mRNA | AK225601.1 | - | 1 - 2 | 91 | 66 | |
AK226059.1 | - | 1 - 2 | 107 | 66 | ||
AK298657.1 | - | 1 - 2 | 93 | 66 | ||
AK298734.1 | - | 1 - 2 | 85 | 66 | ||
BC019931.1 | - | 1 - 2 | 120 | 4536764 | 66 | |
BC020823.1 | - | 1 - 2 | 85 | 4704252 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK298657.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
