Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011514253.2 | XP_011512555.1 | 7 - 8 | 947 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
