Assay Details
| Gene Symbol: GALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138801.2 | NP_620156.1 | 5 - 6 | 1028 | 71 | |
| GenBank mRNA | AK291489.1 | - | 5 - 6 | 836 | 71 | |
| BC014916.1 | - | 5 - 6 | 810 | 3920640 | 71 | |
| BC019263.1 | - | 5 - 6 | 841 | 3502667 | 71 | |
| EU794611.1 | - | 5 - 6 | 810 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000245.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003639.hg.1 | TC02000245.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003639.hg.1 | ENST00000272252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003639.hg.1 | ENST00000410063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003639.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003640.hg.1 | TC02000245.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003640.hg.1 | ENST00000272252.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003640.hg.1 | ENST00000410063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003640.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150490.hg.1 | TC0200007296.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150491.hg.1 | TC0200007296.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000272252.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000410063.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000427858.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000434934.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | ENST00000444351.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007296.hg.1 | NM_138801.2 | Refseq |

GALM
Genomic Map