Assay Details
Gene Symbol: PDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142386.2 | NP_001135858.1 | 6 - 7 | 904 | 94 | |
NM_005391.4 | NP_005382.1 | 6 - 7 | 904 | 94 | ||
GenBank mRNA | AK301965.1 | - | 6 - 7 | 907 | 94 | |
BC015948.1 | - | 6 - 7 | 851 | 3922929 | 94 | |
L42452.1 | - | 6 - 7 | 697 | 94 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000118.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
glucose metabolic process
regulation of acetyl-CoA biosynthetic process from pyruvate
regulation of glucose metabolic process
peptidyl-serine phosphorylation
peroxisome proliferator activated receptor signaling pathway
cellular response to glucose stimulus
cellular response to fatty acid
hypoxia-inducible factor-1alpha signaling pathway
regulation of reactive oxygen species metabolic process
protein kinase activity
protein serine/threonine kinase activity
pyruvate dehydrogenase (acetyl-transferring) kinase activity
protein binding
ATP binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X001587.hg.1 | TC0X000118.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001587.hg.1 | ENST00000379162.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001587.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001587.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X001587.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | ENST00000568479.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00146683.hg.1 | TC0X00006816.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000379162.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000441463.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000493226.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | ENST00000568479.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_001142386.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00006816.hg.1 | NM_005391.4 | Refseq |
