Assay Details
| Gene Symbol: NOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518159.1 | XP_011516461.1 | 2 - 3 | 301 | 55 | |
| XM_011518160.1 | XP_011516462.1 | 2 - 3 | 301 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011518159.1, XM_011518160.1 transcripts.
Set Membership: |
|

NOXA1
Genomic Map