Assay Details
| Gene Symbol: NOXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011518158.2 | XP_011516460.1 | 4 - 5 | 657 | 74 | |
| GenBank mRNA | AK098341.1 | - | 4 - 5 | 482 | 74 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
superoxide metabolic process
regulation of hydrogen peroxide metabolic process
positive regulation of catalytic activity
regulation of respiratory burst
protein binding
superoxide-generating NADPH oxidase activator activity
SH3 domain binding
enzyme binding
Rac GTPase binding
superoxide metabolic process
regulation of hydrogen peroxide metabolic process
positive regulation of catalytic activity
regulation of respiratory burst
protein binding
superoxide-generating NADPH oxidase activator activity
SH3 domain binding
enzyme binding
Rac GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000341349.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | ENST00000392815.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256067.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_001256068.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900009290.hg.1 | NM_006647.1 | Refseq |

NOXA1
Genomic Map