Assay Details
Gene Symbol: B3GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033171.2 | NP_149361.1 | 2 - 3 | 387 | 82 | |
XM_017028231.1 | XP_016883720.1 | 4 - 5 | 4436 | 82 | ||
XM_017028239.1 | XP_016883728.1 | 4 - 5 | 4303 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21000177.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_033171.2,XM_017028231.1,XM_017028239.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | ENST00000343118.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | ENST00000380618.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | ENST00000380620.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR21001719.hg.1 | TC21000177.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | TC2100008521.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100158279.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000343118.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000380618.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000398714.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | ENST00000615480.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_001278650.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_006057.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033170.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033171.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2100008521.hg.1 | NM_033172.2 | Refseq |
