Assay Details
Gene Symbol: HSD17B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016142.2 | NP_057226.1 | 3 - 4 | 508 | 106 | |
XM_011520156.1 | XP_011518458.1 | 3 - 4 | 502 | 106 | ||
XM_017017881.1 | XP_016873370.1 | 3 - 4 | 188 | 106 | ||
GenBank mRNA | AF078850.1 | - | 3 - 4 | 346 | 106 | |
AK074952.1 | - | 3 - 4 | 360 | 106 | ||
AK075216.1 | - | 3 - 4 | 369 | 106 | ||
AK222881.1 | - | 3 - 4 | 330 | 106 | ||
AK292625.1 | - | 3 - 4 | 393 | 106 | ||
AK301663.1 | - | 4 - 5 | 299 | 106 | ||
BC012043.1 | - | 3 - 4 | 331 | 3622879 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000367.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011520156.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
long-chain fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
protein binding
collagen binding
heparin binding
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
3-oxo-arachidoyl-CoA reductase activity
3-oxo-behenoyl-CoA reductase activity
3-oxo-lignoceroyl-CoA reductase activity
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
long-chain fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
protein binding
collagen binding
heparin binding
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
3-oxo-arachidoyl-CoA reductase activity
3-oxo-behenoyl-CoA reductase activity
3-oxo-lignoceroyl-CoA reductase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11004539.hg.1 | ENST00000278353.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004539.hg.1 | ENST00000529261.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11004539.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11004539.hg.1 | TC11000367.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000278353.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000527213.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000531185.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | ENST00000532178.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | NM_016142.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100200200.hg.1 | TC1100012992.hg.1 | Clariom D TC |
