Assay Details
| Gene Symbol: HSD17B12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016142.2 | NP_057226.1 | 5 - 6 | 698 | 124 | |
| XM_011520156.1 | XP_011518458.1 | 5 - 6 | 692 | 124 | ||
| XM_017017881.1 | XP_016873370.1 | 5 - 6 | 378 | 124 | ||
| GenBank mRNA | AF078850.1 | - | 5 - 6 | 536 | 124 | |
| AK027882.1 | - | 591 | 124 | |||
| AK074952.1 | - | 5 - 6 | 550 | 124 | ||
| AK075011.1 | - | 5 - 6 | 583 | 124 | ||
| AK075216.1 | - | 5 - 6 | 559 | 124 | ||
| AK222881.1 | - | 5 - 6 | 520 | 124 | ||
| AK292625.1 | - | 5 - 6 | 583 | 124 | ||
| AK301663.1 | - | 6 - 7 | 489 | 124 | ||
| BC012043.1 | - | 5 - 6 | 521 | 3622879 | 124 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000367.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
long-chain fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
protein binding
collagen binding
heparin binding
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
3-oxo-arachidoyl-CoA reductase activity
3-oxo-behenoyl-CoA reductase activity
3-oxo-lignoceroyl-CoA reductase activity
fatty acid biosynthetic process
steroid biosynthetic process
estrogen biosynthetic process
positive regulation of cell-substrate adhesion
extracellular matrix organization
long-chain fatty-acyl-CoA biosynthetic process
oxidation-reduction process
fibronectin binding
estradiol 17-beta-dehydrogenase activity
protein binding
collagen binding
heparin binding
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
3-oxo-arachidoyl-CoA reductase activity
3-oxo-behenoyl-CoA reductase activity
3-oxo-lignoceroyl-CoA reductase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11004541.hg.1 | ENST00000278353.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004541.hg.1 | ENST00000529261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004541.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11004541.hg.1 | TC11000367.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100200203.hg.1 | TC1100012992.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000278353.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000395700.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000525736.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527213.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000527433.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000531185.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000532178.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533090.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000533802.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | ENST00000534053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012992.hg.1 | NM_016142.2 | Refseq |

HSD17B12
Genomic Map