Assay Details
| Gene Symbol: CASP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080125.1 | NP_001073594.1 | 4 - 5 | 933 | 101 | |
| NM_033355.3 | NP_203519.1 | 5 - 6 | 764 | 101 | ||
| NR_111983.1 | 4 - 5 | 933 | 101 | |||
| XM_005246886.1 | XP_005246943.1 | 4 - 5 | 679 | 101 | ||
| XM_005246887.1 | XP_005246944.1 | 5 - 6 | 831 | 101 | ||
| XM_005246888.1 | XP_005246945.1 | 5 - 6 | 773 | 101 | ||
| XM_005246889.1 | XP_005246946.1 | 4 - 5 | 650 | 101 | ||
| XM_005246890.3 | XP_005246947.1 | 4 - 5 | 657 | 101 | ||
| XM_005246891.4 | XP_005246948.1 | 4 - 5 | 897 | 101 | ||
| XM_006712789.1 | XP_006712852.1 | 5 - 6 | 790 | 101 | ||
| XM_006712790.3 | XP_006712853.1 | 4 - 5 | 669 | 101 | ||
| XM_011511969.1 | XP_011510271.1 | 2 - 3 | 234 | 101 | ||
| GenBank mRNA | AF380342.1 | - | 4 - 5 | 640 | 101 | |
| AF422928.1 | - | 5 - 6 | 752 | 101 | ||
| BC028223.2 | - | 3 - 4 | 582 | 101 | ||
| U58143.1 | - | 3 - 4 | 555 | 101 | ||
| X98172.1 | - | 4 - 5 | 846 | 101 | ||
| X98177.1 | - | 3 - 4 | 555 | 101 | ||
More Information
Set Membership: |
|

CASP8
Genomic Map