Assay Details
| Gene Symbol: C12orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301834.1 | NP_001288763.1 | 3 - 4 | 405 | 85 | |
| NM_001301836.1 | NP_001288765.1 | 2 - 3 | 402 | 85 | ||
| NM_001301838.1 | NP_001288767.1 | 2 - 3 | 630 | 85 | ||
| NM_138425.3 | NP_612434.1 | 2 - 3 | 534 | 85 | ||
| GenBank mRNA | AK311912.1 | - | 2 - 3 | 332 | 85 | |
| BC009925.2 | - | 2 - 3 | 311 | 3537206 | 85 | |
| BE888740.1 | - | 2 - 3 | 321 | 85 | ||
| BP318750.2 | - | 138 | 85 | |||
| CB161655.1 | - | 3 - 4 | 405 | 85 | ||
| DB494917.1 | - | 2 - 3 | 402 | 85 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28607264 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178331 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28607249 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178326 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28607248 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28607262 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178337 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28607257 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178324 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16527634 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P08178332 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 40978 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 8x60K | 5343 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 732498 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P350551 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P350551 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 30552 | - | - |
| Agilent Human Genome CGH Microarray 44B | A_14_P130676 | - | - |
| Agilent Human Genome CGH Microarray 44A | A_14_P130676 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 26921 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P33113241 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P19448339 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_18_P19127381 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P350551 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P350551 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607264 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16527634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178334 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607259 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607244 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607249 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178332 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607248 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607257 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178326 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178337 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178331 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16527634 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08178338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28607264 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P350551 | - | - |
| Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P130676 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178334 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178332 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178329 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178331 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178335 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178325 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178327 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178338 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P08178339 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16527628 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16527631 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16527627 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16527629 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P08178335 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16527630 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P08178334 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P08178332 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P08178331 | - | - |
| Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P08178333 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P130676 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P130676 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P350551 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 113435 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 85173 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_14_P130676 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39574610 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178361 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178387 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178335 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178338 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178329 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178327 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16527634 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178380 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178271 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178323 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178353 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178340 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16527638 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178278 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178394 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178269 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178362 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178381 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178391 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178275 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178384 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178334 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178273 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178382 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178372 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178332 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178370 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178364 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16527631 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P08178360 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16527646 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_14_P130676 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_138425_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7953567 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7953568 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7953565 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7953566 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7953564 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 224719_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715680_PM_s_at | - | - |
| Affymetrix Human Genome U219 Array | 11715680_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224719_PM_s_at | - | - |
| Human Genome U133 Plus 2.0 Array | 224719_s_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_138425_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 224719_s_at6 | - | - |
| Human Genome U95 Set | 46712_at | - | - |
| Human Genome U95 Set | 46712_PM_at | - | - |
| Human Genome U95 Set | 88217_PM_at | - | - |
| Human Genome U95 Set | 88217_at | - | - |
| Human Genome U133 Set | 224719_s_at | - | - |
| Human Genome U133 Set | 224719_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048299 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A048299 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 197519 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34147536-S | NM_138425.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34147536-S | AK311912.1 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34147536-S | BC009925.2 | human_transcriptome |
| Sentrix Human-6 Expression BeadChip | GI_34147536-S | CR620372.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_23505-S | - | - |
| Illumina human-6 v1.0 expression beadchip | 0004610047 | - | - |
| Illumina humanRef-8 v1.0 expression beadchip | 0004610047 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 1170400 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_23505 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1812191 | CR620372.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1812191 | NM_138425.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1812191 | AK311912.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1812191 | BC009925.2 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1812191 | CR620372.1 | human_transcriptome |

C12orf57
Genomic Map