Assay Details
| Gene Symbol: NDUFB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004548.2 | NP_004539.1 | 2 - 3 | 375 | 83 | |
| GenBank mRNA | AF044954.1 | - | 2 - 3 | 339 | 83 | |
| AF067169.1 | - | 2 - 3 | 328 | 83 | ||
| AF088991.1 | - | 2 - 3 | 370 | 83 | ||
| AK291876.1 | - | 2 - 3 | 331 | 83 | ||
| AK311744.1 | - | 2 - 3 | 351 | 83 | ||
| BC005829.2 | - | 2 - 3 | 333 | 2966710 | 83 | |
| BC007509.2 | - | 2 - 3 | 330 | 3030207 | 83 | |
| BP319067.1 | - | 2 - 3 | 383 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000050.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16001118.hg.1 | ENST00000268668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001118.hg.1 | ENST00000543683.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001118.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16001118.hg.1 | TC16000050.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009546.hg.1 | ENST00000567319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009546.hg.1 | ENST00000567319.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146000.hg.1 | TC1600006574.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006574.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009546.hg.1 | ENST00000567319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009546.hg.1 | ENST00000567319.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000268668.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000569148.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | ENST00000570172.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | NM_004548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600146002.hg.1 | TC1600006574.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600146004.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146004.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146004.hg.1 | ENST00000543683.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146004.hg.1 | ENST00000565031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600146004.hg.1 | TC1600006574.hg.1 | Clariom D TC |

NDUFB10
Genomic Map