Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032593.2 | NP_115982.1 | 3 - 4 | 389 | 85 | |
GenBank mRNA | AF085236.1 | - | 3 - 4 | 383 | 85 | |
AF356515.1 | - | 3 - 4 | 353 | 85 | ||
AF356875.1 | - | 3 - 4 | 365 | 85 | ||
AK222582.1 | - | 3 - 4 | 346 | 85 | ||
AY033094.1 | - | 3 - 4 | 360 | 85 | ||
BC047737.2 | - | 3 - 4 | 365 | 6018376 | 85 | |
BI759951.1 | - | 3 - 4 | 389 | 85 | ||
BM764627.1 | - | 3 - 4 | 386 | 85 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001064.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09014248.hg.1 | TC09001064.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09014248.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09014248.hg.1 | ENST00000474908.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09014248.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900168638.hg.1 | TC0900009957.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000259667.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000461169.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000471774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000472085.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474848.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000474908.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | ENST00000490578.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009957.hg.1 | NM_032593.2 | Refseq |
