Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002305.hg.1 |
TC09000201.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002305.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09002305.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
ENST00000616606.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000475774.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000479053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
ENST00000616606.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007113.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148437.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148437.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148437.hg.1 |
ENST00000377966.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900148437.hg.1 |
NM_021111.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900148437.hg.1 |
TC0900007113.hg.1 |
Clariom D TC |