Assay Details
Gene Symbol: RECK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316346.1 | NP_001303275.1 | 2 - 3 | 243 | 104 | |
NM_001316347.1 | NP_001303276.1 | 2 - 3 | 243 | 104 | ||
NM_001316348.1 | NP_001303277.1 | 2 - 3 | 243 | 104 | ||
NM_021111.2 | NP_066934.1 | 2 - 3 | 243 | 104 | ||
XM_017015208.1 | XP_016870697.1 | 2 - 3 | 251 | 104 | ||
XM_017015211.1 | XP_016870700.1 | 2 - 3 | 245 | 104 | ||
GenBank mRNA | AK292653.1 | - | 2 - 3 | 243 | 104 | |
AK315073.1 | - | 2 - 3 | 229 | 104 | ||
BC060806.1 | - | 2 - 3 | 217 | 104 | ||
BC137093.1 | - | 2 - 3 | 204 | 9021090 | 104 | |
BX393623.2 | - | 2 - 3 | 231 | 104 | ||
D50406.1 | - | 2 - 3 | 266 | 104 | ||
JQ756320.1 | - | 2 - 3 | 157 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000201.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017015208.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel maturation
embryo implantation
extracellular matrix organization
negative regulation of cell migration
embryonic forelimb morphogenesis
negative regulation of metalloendopeptidase activity
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
protein binding
metalloendopeptidase inhibitor activity
blood vessel maturation
embryo implantation
extracellular matrix organization
negative regulation of cell migration
embryonic forelimb morphogenesis
negative regulation of metalloendopeptidase activity
endopeptidase inhibitor activity
serine-type endopeptidase inhibitor activity
protein binding
metalloendopeptidase inhibitor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR09002279.hg.1 | TC09000201.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002279.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002279.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002279.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002282.hg.1 | TC09000201.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002282.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002282.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002282.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | ENST00000616606.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09002280.hg.1 | TC09000201.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09002280.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002280.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09002280.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148415.hg.1 | TC0900007113.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | ENST00000616606.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007113.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148412.hg.1 | TC0900007113.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148413.hg.1 | TC0900007113.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000377966.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000475774.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | ENST00000479053.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | NM_021111.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900148411.hg.1 | TC0900007113.hg.1 | Clariom D TC |
