Assay Details
| Gene Symbol: RGS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286486.1 | NP_001273415.1 | 1 - 2 | 107 | 88 | |
| NM_003834.2 | NP_003825.1 | 1 - 2 | 107 | 88 | ||
| XM_011522720.2 | XP_011521022.1 | 1 - 2 | 34 | 88 | ||
| GenBank mRNA | AF035154.1 | - | 1 - 2 | 78 | 88 | |
| AK091701.1 | - | 1 - 2 | 104 | 88 | ||
| AK308972.1 | - | 1 - 2 | 32 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF035154.1, NM_001286486.1, NM_003834.2, XM_011522720.2 transcripts.
Set Membership: |
|

RGS11
Genomic Map