Assay Details
Gene Symbol: TLR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003268.5 | NP_003259.2 | 4 - 5 | 471 | 100 | |
XM_005273243.4 | XP_005273300.2 | 3 - 4 | 323 | 100 | ||
XM_006711505.3 | XP_006711568.1 | 2 - 3 | 268 | 100 | ||
XM_017002208.1 | XP_016857697.1 | 3 - 4 | 384 | 100 | ||
GenBank mRNA | DB448079.1 | - | 3 - 4 | 348 | 100 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003871.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_003268.5,XM_005273243.4,XM_006711505.3,XM_017002208.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
male gonad development
positive regulation of interleukin-8 production
positive regulation of toll-like receptor signaling pathway
toll-like receptor 5 signaling pathway
innate immune response
positive regulation of nitric oxide biosynthetic process
regulation of cytokine secretion
defense response to Gram-negative bacterium
cellular response to lipopolysaccharide
cellular response to mechanical stimulus
receptor activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
MyD88-dependent toll-like receptor signaling pathway
inflammatory response
male gonad development
positive regulation of interleukin-8 production
positive regulation of toll-like receptor signaling pathway
toll-like receptor 5 signaling pathway
innate immune response
positive regulation of nitric oxide biosynthetic process
regulation of cytokine secretion
defense response to Gram-negative bacterium
cellular response to lipopolysaccharide
cellular response to mechanical stimulus
receptor activity
transmembrane signaling receptor activity
interleukin-1 receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | TC01003871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | ENST00000342210.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | ENST00000366881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | ENST00000540964.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058868.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01058865.hg.1 | TC01003871.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01058865.hg.1 | ENST00000342210.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058865.hg.1 | ENST00000366881.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058865.hg.1 | ENST00000540964.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01058865.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000465726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100017445.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000484766.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231863.hg.1 | TC0100017445.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000366881.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000407096.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | ENST00000540964.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | NM_003268.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100231860.hg.1 | TC0100017445.hg.1 | Clariom D TC |
