Assay Details
| Gene Symbol: MAOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000240.3 | NP_000231.1 | 12 - 13 | 2701 | 72 | |
| NM_001270458.1 | NP_001257387.1 | 13 - 14 | 2809 | 72 | ||
| GenBank mRNA | AK223499.1 | - | 12 - 13 | 1443 | 72 | |
| AK291769.1 | - | 12 - 13 | 1305 | 72 | ||
| AK293926.1 | - | 13 - 14 | 1416 | 72 | ||
| BC008064.2 | - | 12 - 13 | 1287 | 2990003 | 72 | |
| BC044787.1 | - | 12 - 13 | 2703 | 4824582 | 72 | |
| BT006651.1 | - | 12 - 13 | 1266 | 72 | ||
| M68840.1 | - | 12 - 13 | 1339 | 72 | ||
| M69226.1 | - | 12 - 13 | 1316 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cellular biogenic amine metabolic process
neurotransmitter metabolic process
neurotransmitter catabolic process
dopamine catabolic process
serotonin metabolic process
phenylethylamine metabolic process
oxidation-reduction process
primary amine oxidase activity
flavin adenine dinucleotide binding
serotonin binding
cellular biogenic amine metabolic process
neurotransmitter metabolic process
neurotransmitter catabolic process
dopamine catabolic process
serotonin metabolic process
phenylethylamine metabolic process
oxidation-reduction process
primary amine oxidase activity
flavin adenine dinucleotide binding
serotonin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00148024.hg.1 | TC0X00007053.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000338702.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000490604.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000497485.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | ENST00000542639.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_000240.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00007053.hg.1 | NM_001270458.1 | Refseq |

MAOA
Genomic Map