Assay Details
| Gene Symbol: EIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015904.3 | NP_056988.3 | 7 - 8 | 1571 | 115 | |
| GenBank mRNA | AB018284.1 | - | 7 - 8 | 1537 | 115 | |
| AF078035.1 | - | 7 - 8 | 1504 | 115 | ||
| AJ006776.1 | - | 7 - 8 | 1527 | 115 | ||
| AK308176.1 | - | 7 - 8 | 1553 | 115 | ||
| AL133563.1 | - | 4 - 5 | 1123 | 115 | ||
| BC032639.1 | - | 7 - 8 | 1493 | 5496504 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000603.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009543.hg.1 | TC02000603.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009543.hg.1 | ENST00000289371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02009543.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200160003.hg.1 | TC0200008587.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000289371.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000470868.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000494190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | ENST00000617677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008587.hg.1 | NM_015904.3 | Refseq |

EIF5B
Genomic Map