Assay Details
Gene Symbol: KIF4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012310.4 | NP_036442.3 | 4 - 5 | 564 | 166 | |
GenBank mRNA | AB208797.1 | - | 4 - 5 | 464 | 166 | |
AF071592.2 | - | 4 - 5 | 481 | 166 | ||
AF179308.1 | - | 3 - 4 | 421 | 166 | ||
AJ271784.1 | - | 3 - 4 | 427 | 166 | ||
AK313133.1 | - | 3 - 4 | 797 | 166 | ||
BC003664.2 | - | 4 - 5 | 502 | 166 | ||
BC038459.1 | - | 4 - 5 | 501 | 166 | ||
BC049218.1 | - | 4 - 5 | 470 | 166 | ||
BC050548.1 | - | 4 - 5 | 463 | 4538604 | 166 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000376.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
retrograde vesicle-mediated transport, Golgi to ER
organelle organization
microtubule-based movement
anterograde axonal transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
mitotic spindle midzone assembly
DNA binding
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
mitotic cytokinesis
retrograde vesicle-mediated transport, Golgi to ER
organelle organization
microtubule-based movement
anterograde axonal transport
antigen processing and presentation of exogenous peptide antigen via MHC class II
mitotic spindle midzone assembly
DNA binding
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004854.hg.1 | TC0X000376.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004854.hg.1 | ENST00000374388.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004854.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X004854.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151636.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151636.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151636.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00151636.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00151636.hg.1 | TC0X00007537.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000374403.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | ENST00000485406.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007537.hg.1 | NM_012310.4 | Refseq |
